accessibility menu, dialog, popup

UserName

CAS RN 3581-89-3

IUPAC Name: 5-methyl-1,3-thiazole
Synonyms: 5-methyl-1,3-thiazole
Molecular Weight (g/mol): 99.15
Molecular Formula: C4H5NS
InChi Key: RLYUNPNLXMSXAX-UHFFFAOYSA-N
SMILES: CC1=CN=CS1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)