CAS RN 35779-04-5
IUPAC Name:
1-tert-butyl-4-iodobenzene
Synonyms:
1-tert-butyl-4-iodobenzene
Molecular Weight (g/mol):
260.12
Molecular Formula:
C10H13I
InChi Key:
WQVIVQDHNKQWTM-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(I)C=C1
1
–
1
of
1
results
1
1-tert-Butyl-4-iodobenzene, 97%
CAS: 35779-04-5 Molecular Formula: C10H13I Molecular Weight (g/mol): 260.118 MDL Number: MFCD00052339 InChI Key: WQVIVQDHNKQWTM-UHFFFAOYSA-N Synonym: 1-tert-butyl-4-iodobenzene,4-tert-butyliodobenzene,4-t-butyliodobenzene,1-iodo-4-tert-butylbenzene,benzene, 1-1,1-dimethylethyl-4-iodo,4-tert-butyl-1-iodobenzene,2-4-iodophenyl-2-methylpropane,2-4'-iodophenyl-2-methylpropane,pubchem3974,4-t-butyl-iodobenzene PubChem CID: 142029 IUPAC Name: 1-tert-butyl-4-iodobenzene SMILES: CC(C)(C)C1=CC=C(C=C1)I