accessibility menu, dialog, popup

UserName

CAS RN 35405-71-1

OOOHOHOOHOOHOHHO

CAS RN 35405-71-1

IUPAC Name: 2-({3,4-dihydroxy-6,8-dioxabicyclo[3.2.1]octan-2-yl}oxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-({3,4-dihydroxy-6,8-dioxabicyclo[3.2.1]octan-2-yl}oxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 324.28
Molecular Formula: C12H20O10
InChi Key: LTYZUJSCZCPGHH-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C3COC(O3)C(O)C2O)C(O)C(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)