accessibility menu, dialog, popup

UserName

OHClCl

CAS RN 35364-79-5

IUPAC Name: 2-(3,4-dichlorophenyl)ethan-1-ol
Synonyms: 2-(3,4-dichlorophenyl)ethanol
Molecular Weight (g/mol): 191.05
Molecular Formula: C8H8Cl2O
InChi Key: GITOMJDYNUMCOV-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results