CAS RN 35243-43-7
IUPAC Name:
2-ethoxy-3-ethylpyrazine
Synonyms:
2-ethoxy-3-ethylpyrazine
Molecular Weight (g/mol):
152.2
Molecular Formula:
C8H12N2O
InChi Key:
TYSHGGVMHVIOMH-UHFFFAOYSA-N
SMILES:
CCOC1=NC=CN=C1CC
1
–
1
of
1
results
1
2-Ethoxy-3-ethylpyrazine, TRC
CAS: 35243-43-7 Molecular Formula: C8H12N2O Molecular Weight (g/mol): 152.19 SMILES: CCOc1nccnc1CC