CAS RN 35219-13-7
IUPAC Name:
4-(1,3,4-oxadiazol-2-yl)aniline
Synonyms:
4-(1,3,4-oxadiazol-2-yl)aniline
Molecular Weight (g/mol):
161.16
Molecular Formula:
C8H7N3O
InChi Key:
WKERLKMCAVNWGD-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)C1=NN=CO1
1
–
1
of
1
results
1
4-(1,3,4-Oxadiazol-2-yl)aniline, TRC
CAS: 35219-13-7 Molecular Formula: C8H7N3O Molecular Weight (g/mol): 161.16 IUPAC Name: 4-(1,3,4-oxadiazol-2-yl)aniline SMILES: Nc1ccc(cc1)c2ocnn2