accessibility menu, dialog, popup

UserName

CAS RN 351-04-2

FHNOCl

CAS RN 351-04-2

IUPAC Name: 2-chloro-N-(4-fluorophenyl)acetamide
Synonyms: 2-chloro-N-(4-fluorophenyl)acetamide
Molecular Weight (g/mol): 187.6
Molecular Formula: C8H7ClFNO
InChi Key: JDAWWCJBFPBHFL-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC(=O)CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results