CAS RN 35065-30-6
IUPAC Name:
2,2',3,3',4,4',5-heptachloro-1,1'-biphenyl
Synonyms:
2,2',3,3',4,4',5-heptachloro-1,1'-biphenyl
Molecular Weight (g/mol):
395.31
Molecular Formula:
C12H3Cl7
InChi Key:
RMPWIIKNWPVWNG-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C(Cl)=C1Cl)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
2,2',3,3',4,4',5-Heptachloro-1,1'-biphenyl, TRC
CAS: 35065-30-6 Molecular Formula: C12 H3 Cl7 Molecular Weight (g/mol): 395.32 Synonym: 1,1'-Biphenyl, 2,2',3,3',4,4',5-heptachloro-,2,2',3,3',4,4',5-Heptachloro-1,1'-biphenyl,2,2',3,3',4,4',5-Heptachlorobiphenyl,2,3,4,2',3',4',5'-Heptachlorobiphenyl,2,3,4,5,2',3',4'-Heptachlorobiphenyl,CB 170,PCB 170,2,2',3,3',4,4',5-HeptaCB IUPAC Name: 1,2,3,4-tetrachloro-5-(2,3,4-trichlorophenyl)benzene SMILES: Clc1ccc(c(Cl)c1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl