accessibility menu, dialog, popup

UserName

ClClClClClCl

CAS RN 35065-27-1

IUPAC Name: 2,2',4,4',5,5'-hexachloro-1,1'-biphenyl
Synonyms: hexachlorobiphenyl HCBP
Molecular Weight (g/mol): 360.86
Molecular Formula: C12H4Cl6
InChi Key: MVWHGTYKUMDIHL-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C=C1Cl)C1=CC(Cl)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (2)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results