accessibility menu, dialog, popup

UserName

CH3CH3OCH3CH3OCH3CH3OSm

CAS RN 3504-40-3

IUPAC Name: samarium(3+) tris(propan-2-olate)
Synonyms: samarium(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 327.62
Molecular Formula: C9H21O3Sm
InChi Key: HJCRVWSKQNDSPZ-UHFFFAOYSA-N
SMILES: [Sm+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results