CAS RN 35022-72-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diquat Dipyridone, TRC
CAS: 35022-72-1 Molecular Formula: C12 H10 N2 O2 Molecular Weight (g/mol): 214.22 Synonym: 6,7-Dihydro-dipyrido[1,2-a:2',1'-c]pyrazine-4,9-dione IUPAC Name: 7,10-diazatricyclo[8.4.0.02,7]tetradeca-1(14),2,4,12-tetraene-6,11-dione SMILES: O=C1C=CC=C2N1CCN3C(=O)C=CC=C23