accessibility menu, dialog, popup

UserName

CAS RN 34576-85-7

ClSClOCl

CAS RN 34576-85-7

IUPAC Name: 3,6-dichloro-1-benzothiophene-2-carbonyl chloride
Synonyms: 3,6-dichloro-1-benzothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 265.53
Molecular Formula: C9H3Cl3OS
InChi Key: JCTLRFNZVZCAHR-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)C2=CC=C(Cl)C=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results