accessibility menu, dialog, popup

UserName

CH3NH2H3CN

CAS RN 3430-33-9

IUPAC Name: 2,6-dimethylpyridin-3-amine
Synonyms: 2,6-dimethylpyridin-3-amine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: WISXXOGOMDYNSN-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(C)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results