accessibility menu, dialog, popup

UserName

H3COSBrOCH3

CAS RN 34042-95-0

IUPAC Name: 3-bromo-2-(diethoxymethyl)thiophene
Synonyms: 3-bromo-2-(diethoxymethyl)thiophene
Molecular Weight (g/mol): 265.17
Molecular Formula: C9H13BrO2S
InChi Key: QNTDBERHEOAJAA-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=C(Br)C=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results