CAS RN 340256-40-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Ethylphenol-2,3,5,6-d4,OD, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
4-Ethylphenol-2,3,5,6-d4,OD, TRC
CAS: 340256-40-8 Molecular Formula: C8 D5 H5 O Molecular Weight (g/mol): 127.1952 Synonym: Phen-2,3,5,6-d4-ol-d, 4-ethyl- (9CI) IUPAC Name: 1,2,4,5-tetradeuterio-3-deuteriooxy-6-ethylbenzene SMILES: [2H]Oc1c([2H])c([2H])c(CC)c([2H])c1[2H]