CAS RN 33967-19-0
CAS RN 33967-19-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(3,4-Dimethylphenyl)ethanol, TRC
CAS: 33967-19-0 Molecular Formula: C10 H14 O Molecular Weight (g/mol): 150.22 Synonym: Medetomidine Impurity 44 IUPAC Name: 1-(3,4-dimethylphenyl)ethanol SMILES: CC(O)c1ccc(C)c(C)c1