accessibility menu, dialog, popup

UserName

CH3OH(E)(E)

CAS RN 33467-79-7

IUPAC Name: (2E,4E)-hepta-2,4-dien-1-ol
Synonyms: (2E,4E)-hepta-2,4-dien-1-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: MDRZSADXFOPYOC-VNKDHWASSA-N
SMILES: CC\C=C\C=C\CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results