CAS RN 334512-82-2
IUPAC Name:
zirconium(4+) calcium hexakis(propan-2-olate)
Synonyms:
zirconium(4+) calcium hexakis(propan-2-olate)
Molecular Weight (g/mol):
485.83
Molecular Formula:
C18H42CaO6Zr
InChi Key:
VGAKIAWXLKTFPJ-UHFFFAOYSA-N
SMILES:
[Ca++].[Zr+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
1
–
1
of
1
results
1
Calcium zirconium isopropoxide, 10% w/v in isopropanol, Thermo Scientific™
CAS: 334512-82-2 Molecular Formula: C18H42CaO6Zr Molecular Weight (g/mol): 485.83 MDL Number: MFCD01865977 InChI Key: VGAKIAWXLKTFPJ-UHFFFAOYSA-N Synonym: zirconium 4+ calcium hexakis propan-2-olate,calcium zirconium isopropoxide w/v in isopropanol PubChem CID: 102602388 IUPAC Name: calcium;propan-2-olate;zirconium(4+) SMILES: CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ca+2].[Zr+4]