CAS RN 331947-44-5
CAS RN 331947-44-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(-)-Tolvaptan, TRC
CAS: 331947-44-5 Molecular Formula: C26 H25 Cl N2 O3 Molecular Weight (g/mol): 448.94 Synonym: N-[4-[[(5S)-7-Chloro-2,3,4,5-tetrahydro-5-hydroxy-1H-1-benzazepin-1-yl]carbonyl]-3-methylphenyl]-2-methylbenzamide,(S)-(-)-OPC 41061 IUPAC Name: N-[4-[(5R)-7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl]-3-methylphenyl]-2-methylbenzamide SMILES: Cc1ccccc1C(=O)Nc2ccc(C(=O)N3CCC[C@@H](O)c4cc(Cl)ccc34)c(C)c2