accessibility menu, dialog, popup

UserName

CAS RN 33184-16-6

CH3FOHO

CAS RN 33184-16-6

IUPAC Name: 5-fluoro-2-methylbenzoic acid
Synonyms: 5-fluoro-2-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: JVBLXLBINTYFPR-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results