accessibility menu, dialog, popup

UserName

CAS RN 331761-46-7

IUPAC Name: 4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Synonyms: 4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Molecular Weight (g/mol): 281.15
Molecular Formula: C12H6Cl2N2S
InChi Key: QDMQHIKVIOWCGY-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CSC2=NC=NC(Cl)=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results