CAS RN 329940-70-7
IUPAC Name:
2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
Synonyms:
2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
Molecular Weight (g/mol):
290.7
Molecular Formula:
C14H11ClN2O3
InChi Key:
VIQSGUZRECTJEM-UHFFFAOYSA-N
SMILES:
CC1=CC(NC(=O)C2=C(Cl)C=C(C=C2)[N+]([O-])=O)=CC=C1
1
–
1
of
1
results
1
2-Chloro-N-(3-methylphenyl)-4-nitrobenzamide, 97%, Thermo Scientific™
CAS: 329940-70-7 Molecular Formula: C14H11ClN2O3 Molecular Weight (g/mol): 290.703 MDL Number: MFCD00784494 InChI Key: VIQSGUZRECTJEM-UHFFFAOYSA-N Synonym: 2-chloro-n-3-methylphenyl-4-nitrobenzamide,2-chloro-4-nitro-n-3-methylphenyl benzamide,cambridge id 5344357,2-chloro-4-nitrophenyl-n-3-methylphenyl carboxamide PubChem CID: 710885 IUPAC Name: 2-chloro-N-(3-methylphenyl)-4-nitrobenzamide SMILES: CC1=CC(=CC=C1)NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl