accessibility menu, dialog, popup

UserName

CH3OHNONOOCl

CAS RN 328258-90-8

IUPAC Name: 2-chloro-N-(4-ethoxyphenyl)-4-nitrobenzamide
Synonyms: 2-chloro-N-(4-ethoxyphenyl)-4-nitrobenzamide
Molecular Weight (g/mol): 320.73
Molecular Formula: C15H13ClN2O4
InChi Key: WKNXXIWQPIKWEA-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(=O)C2=C(Cl)C=C(C=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results