accessibility menu, dialog, popup

UserName

H3COOCH3H2N

CAS RN 3275-95-4

IUPAC Name: 1-(2,5-dimethoxyphenyl)methanamine
Synonyms: 1-(2,5-dimethoxyphenyl)methanamine
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: UWMCHDDHXMFKMA-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(CN)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results