CAS RN 32676-16-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Dihydroperillic Acid, TRC
CAS: 32676-16-7 Molecular Formula: C10H16O2 Molecular Weight (g/mol): 168.23 Synonym: 4-(1-Methylethenyl)cyclohexanecarboxylic Acid SMILES: CC(=C)C1CCC(CC1)C(=O)O