accessibility menu, dialog, popup

UserName

H3CNNCl

CAS RN 32601-86-8

IUPAC Name: 2-chloro-3-methylquinoxaline
Synonyms: 2-chloro-3-methylquinoxaline
Molecular Weight (g/mol): 178.62
Molecular Formula: C9H7ClN2
InChi Key: PXDLUYLWPJMGJA-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results