accessibility menu, dialog, popup

UserName

CAS RN 325145-35-5

CH3(S)NHNOOH3CCH3H3C

CAS RN 325145-35-5

IUPAC Name: tert-butyl (2S)-2-ethylpiperazine-1-carboxylate
Synonyms: tert-butyl (2S)-2-ethylpiperazine-1-carboxylate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: CTCGRXDGXGUOTE-VIFPVBQESA-N
SMILES: CC[C@H]1CNCCN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results