accessibility menu, dialog, popup

UserName

CAS RN 324750-04-1

H3C(S)NHN

CAS RN 324750-04-1

IUPAC Name: (3S)-1-benzyl-3-ethylpiperazine
Synonyms: (3S)-1-benzyl-3-ethylpiperazine
Molecular Weight (g/mol): 204.32
Molecular Formula: C13H20N2
InChi Key: CTPKPBTULPZITK-ZDUSSCGKSA-N
SMILES: CC[C@H]1CN(CC2=CC=CC=C2)CCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results