accessibility menu, dialog, popup

UserName

NON

CAS RN 3237-22-7

IUPAC Name: 2-[(furan-2-yl)methylidene]propanedinitrile
Synonyms: 2-(furan-2-ylmethylidene)propanedinitrile
Molecular Weight (g/mol): 144.13
Molecular Formula: C8H4N2O
InChi Key: CRUJHLHRJSNZIU-UHFFFAOYSA-N
SMILES: N#CC(=CC1=CC=CO1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results