accessibility menu, dialog, popup

UserName

CAS RN 3236-71-3

OHHO

CAS RN 3236-71-3

IUPAC Name: 4-[9-(4-hydroxyphenyl)-9H-fluoren-9-yl]phenol
Synonyms: 4-[9-(4-hydroxyphenyl)fluoren-9-yl]phenol
Molecular Weight (g/mol): 350.42
Molecular Formula: C25H18O2
InChi Key: YWFPGFJLYRKYJZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results