accessibility menu, dialog, popup

UserName

CAS RN 3236-48-4

HOOH

CAS RN 3236-48-4

IUPAC Name: [4-(hydroxymethyl)cyclohexyl]methanol
Synonyms: cyclohexanedimethanol 1,4-cyclohexanedimethanol
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: YIMQCDZDWXUDCA-UHFFFAOYSA-N
SMILES: OCC1CCC(CO)CC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)