accessibility menu, dialog, popup

UserName

CAS RN 3225-26-1

H3CNOH3CH3COOCH3

CAS RN 3225-26-1

IUPAC Name: [4-(benzoyloxy)-2,2,6,6-tetramethylpiperidin-1-yl]oxidanyl
Synonyms: 4-(benzoyloxy)-2,2,6,6-tetramethylpiperidin-1-yloxidanyl
Molecular Weight (g/mol): 276.36
Molecular Formula: C16H22NO3
InChi Key: MJEDTBDGYVATPI-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1[O])OC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)