accessibility menu, dialog, popup

UserName

OSOOClO

CAS RN 321309-38-0

IUPAC Name: 3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride
Synonyms: 3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride
Molecular Weight (g/mol): 248.68
Molecular Formula: C9H9ClO4S
InChi Key: ADAGPISZFDLGCR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(OCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results