CAS RN 31928-47-9
CAS RN 31928-47-9
IUPAC Name:
4-bromo-2-chloro-1-iodobenzene
Synonyms:
4-bromo-2-chloro-1-iodobenzene
Molecular Weight (g/mol):
317.35
Molecular Formula:
C6H3BrClI
InChi Key:
OHHKQBZOURGNLR-UHFFFAOYSA-N
SMILES:
ClC1=C(I)C=CC(Br)=C1
1
–
1
of
1
results
1
4-Bromo-2-chloro-1-iodobenzene, 97%
CAS: 31928-47-9 Molecular Formula: C6H3BrClI Molecular Weight (g/mol): 317.348 MDL Number: MFCD00079709 InChI Key: OHHKQBZOURGNLR-UHFFFAOYSA-N Synonym: 4-bromo-2-chloroiodobenzene,1-bromo-3-chloro-4-iodobenzene,benzene, 4-bromo-2-chloro-1-iodo,buttpark 34\07-78,4-bromo-2-chloro-1-iodo-benzene,pubchem3592,acmc-1cp6y,3-chloro-4-iodobromobenzene,ksc493m0p,4-bromo-2-chloro-iodobenzene PubChem CID: 2735547 IUPAC Name: 4-bromo-2-chloro-1-iodobenzene SMILES: C1=CC(=C(C=C1Br)Cl)I