accessibility menu, dialog, popup

UserName

CAS RN 318280-69-2

NOHOOCl

CAS RN 318280-69-2

IUPAC Name: 1-(2-chloroacetyl)piperidine-4-carboxylic acid
Synonyms: 1-(2-chloroacetyl)piperidine-4-carboxylic acid
Molecular Weight (g/mol): 205.64
Molecular Formula: C8H12ClNO3
InChi Key: PEQVHRULPWCLOJ-UHFFFAOYSA-N
SMILES: OC(=O)C1CCN(CC1)C(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results