accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 31699-38-4

IUPAC Name: dodeca-2,10-diyne
Synonyms: 2,10-dodecadiyne
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: CNMHMGNWEVHOCK-UHFFFAOYSA-N
SMILES: CC#CCCCCCCC#CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results