accessibility menu, dialog, popup

UserName

CAS RN 313657-75-9

CH3NHN

CAS RN 313657-75-9

IUPAC Name: 3-methyl-1-(triphenylmethyl)piperazine
Synonyms: 3-methyl-1-(triphenylmethyl)piperazine
Molecular Weight (g/mol): 342.49
Molecular Formula: C24H26N2
InChi Key: VYUQPZHRDKLOPX-UHFFFAOYNA-N
SMILES: CC1CN(CCN1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results