accessibility menu, dialog, popup

UserName

CH3SONSHNH2C

CAS RN 307512-22-7

IUPAC Name: 11-methyl-4-(prop-2-en-1-yl)-5-sulfanylidene-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7)-dien-3-one
Synonyms: 11-methyl-4-(prop-2-en-1-yl)-5-sulfanylidene-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7)-dien-3-one
Molecular Weight (g/mol): 292.42
Molecular Formula: C14H16N2OS2
InChi Key: BQGPJUIDDGGYJB-UHFFFAOYNA-N
SMILES: CC1CCC2=C(C1)SC1=C2C(=O)N(CC=C)C(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results