accessibility menu, dialog, popup

UserName

CAS RN 305808-14-4

OSOOONa

CAS RN 305808-14-4

IUPAC Name: 2-formylbenzene-1-sulfonic acid hydrate sodium
Synonyms: 2-formylbenzenesulfonic acid hydrate sodium
Molecular Weight (g/mol): 227.19
Molecular Formula: C7H8NaO5S
InChi Key: LYIJOFWETPXEGE-UHFFFAOYSA-N
SMILES: O.[Na].OS(=O)(=O)C1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results