accessibility menu, dialog, popup

UserName

H3CBrH2N

CAS RN 30273-22-4

IUPAC Name: 4-bromo-2-ethylaniline
Synonyms: 4-bromo-2-ethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: LGOZNQPHTIGMQJ-UHFFFAOYSA-N
SMILES: CCC1=CC(Br)=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results