CAS RN 300690-74-8
CAS RN 300690-74-8
IUPAC Name:
3-methylcinnolin-5-amine
Synonyms:
3-methylcinnolin-5-amine
Molecular Weight (g/mol):
159.19
Molecular Formula:
C9H9N3
InChi Key:
HOJQDQQMHMVETF-UHFFFAOYSA-N
SMILES:
CC1=NN=C2C=CC=C(N)C2=C1
1
–
1
of
1
results
1
3-Methylcinnolin-5-amine, 97%, Thermo Scientific™
CAS: 300690-74-8 Molecular Formula: C9H9N3 Molecular Weight (g/mol): 159.19 MDL Number: MFCD00466524 InChI Key: HOJQDQQMHMVETF-UHFFFAOYSA-N PubChem CID: 5310645 IUPAC Name: 3-methylcinnolin-5-amine SMILES: CC1=NN=C2C=CC=C(N)C2=C1