accessibility menu, dialog, popup

UserName

CAS RN 300345-76-0

H3CSOOHNH2N

CAS RN 300345-76-0

IUPAC Name: N-(2-amino-1,2-diphenylethyl)methanesulfonamide
Synonyms: N-(2-amino-1,2-diphenylethyl)methanesulfonamide
Molecular Weight (g/mol): 290.38
Molecular Formula: C15H18N2O2S
InChi Key: FSRRNSLQEDUDTP-UHFFFAOYNA-N
SMILES: CS(=O)(=O)NC(C(N)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results