accessibility menu, dialog, popup

UserName

CAS RN 292636-09-0

FHNBr

CAS RN 292636-09-0

IUPAC Name: 7-bromo-4-fluoro-1H-indole
Synonyms: 7-bromo-4-fluoro-1H-indole
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: PRXUTIXCOBFMMM-UHFFFAOYSA-N
SMILES: FC1=C2C=CNC2=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results