accessibility menu, dialog, popup

UserName

CAS RN 28989-52-8

OSNNH2

CAS RN 28989-52-8

IUPAC Name: 4-(furan-2-yl)-1,3-thiazol-2-amine
Synonyms: 4-(furan-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 166.2
Molecular Formula: C7H6N2OS
InChi Key: CGJGALUSBPYALD-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results