accessibility menu, dialog, popup

UserName

CAS RN 287172-80-9

IUPAC Name: 2-chloro-3,6-difluoroaniline
Synonyms: 2-chloro-3,6-difluoroaniline
Molecular Weight (g/mol): 163.55
Molecular Formula: C6H4ClF2N
InChi Key: LRONWWATBDXKNX-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results