accessibility menu, dialog, popup

UserName

CH3OONOH3CNOCl

CAS RN 2854-32-2

IUPAC Name: 2-[2-(4-chlorobenzoyl)-5-methoxy-2-methyl-2H-indol-3-yl]-1-(morpholin-4-yl)ethan-1-one
Synonyms: 2-[2-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]-1-(morpholin-4-yl)ethanone
Molecular Weight (g/mol): 426.9
Molecular Formula: C23H23ClN2O4
InChi Key: OOKFHWWAUUITQR-UHFFFAOYNA-N
SMILES: COC1=CC2=C(CC(=O)N3CCOCC3)C(C)(N=C2C=C1)C(=O)C1=CC=C(Cl)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results