accessibility menu, dialog, popup

UserName

CAS RN 2842-38-8

HONH

CAS RN 2842-38-8

IUPAC Name: 2-(cyclohexylamino)ethan-1-ol
Synonyms: ethanol, 2-(cyclohexylamino)-
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: MGUMZJAQENFQKN-UHFFFAOYSA-N
SMILES: OCCNC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results