CAS RN 2794-60-7
IUPAC Name:
barium(2+) ditrifluoromethanesulfonate
Synonyms:
trifluoromethanesulfonic acid
Molecular Weight (g/mol):
435.45
Molecular Formula:
C2BaF6O6S2
InChi Key:
DXJURUJRANOYMX-UHFFFAOYSA-L
SMILES:
[Ba++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Barium trifluoromethanesulfonate, Thermo Scientific™
CAS: 2794-60-7 Molecular Formula: C2BaF6O6S2 Molecular Weight (g/mol): 435.453 MDL Number: MFCD00143644 InChI Key: DXJURUJRANOYMX-UHFFFAOYSA-L Synonym: barium trifluoromethanesulfonate,barium triflate,methanesulfonic acid, trifluoro-, barium salt,barium trifluoromethanesulphonate,methanesulfonic acid, 1,1,1-trifluoro-, barium salt 2:1,bariumtrifluoromethanesulfonate,pubchem23759,ksc491a8b,barium 2+ ; trifluoromethanesulfonate PubChem CID: 76040 IUPAC Name: barium(2+);trifluoromethanesulfonate SMILES: C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ba+2]