CAS RN 2708280-12-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
n-Heptyl 4-hydroxybenzoate-d4, TRC
CAS: 2708280-12-8 Molecular Formula: C14 D4 H16 O3 Molecular Weight (g/mol): 240.331 Synonym: n-Heptyl 4-hydroxybenzoate-2,3,5,6-d4,Heptyl alcohol p-hydroxybenzoate-d4,Heptyl 4-hydroxybenzoate-2,3,5,6-D4,Heptyl p-hydroxybenzoate-d4,Heptyl paraben-d4,4-Hydroxybenzoic-2,3,5,6-d4 acid heptyl ester IUPAC Name: heptyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate SMILES: [2H]c1c([2H])c(C(=O)OCCCCCCC)c([2H])c([2H])c1O
2
n-Heptyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.