CAS RN 26787-56-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Distearoyl-2-chloropropanediol, TRC
CAS: 26787-56-4 Molecular Formula: C39 H75 Cl O4 Molecular Weight (g/mol): 643.46 Synonym: Octadecanoic acid, 2-chloro-1,3-propanediyl ester (9CI),Stearic acid, 2-chlorotrimethylene ester (8CI),1,3-Propanediol, 2-chloro-, distearate (8CI),Octadecanoic acid, 2-chloro-1,3-propanediyl ester (9CI),Stearic acid, 2-chlorotrimethylene ester (8CI),1,3-Propanediol, 2-chloro-, distearate (8CI),1,3-Bis-stearoyl-2-chloropropanediol IUPAC Name: (2-chloro-3-octadecanoyloxypropyl) octadecanoate SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(Cl)COC(=O)CCCCCCCCCCCCCCCCC